(2R,6S)-(+)-4-Benzoyloxy-2-benzoyloxymethyl-6-methoxy-2H-pyran-3(6H)-one för syntes
Catalog # 8.18233.0001
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:124 - 127 °C
- Molar Mass:382.37 g/mol
- Synonyms:Methyl-3,6-dibenzoyl-2-deoxy-α-D-glycero-hex-2-enopyranosid-4-ulose
- Hill Formula:C21H18O7
- Storage:+15...+25 °C
- Assay (HPLC, area%):≥99,0% (a/a)
- Specific rotation:70,0 - 71,0 °
- Identity (IR):Passes test
Specification Test Results
About this item
:
:382.368 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:25552-06-1
Formula:C₂₁H₁₈O₇