Blades for safety knives, No. 199
Katalog # 223-0114
Leverantör: MARTOR ARGENTAX
Specifikationer
- Beskrivning:Spare trapezoid blade (no. 199), two-sided deep edge
- L×B:60,0×19 mm
- Material:Rostfritt stål
- Tjocklek:0,63 mm
- Förp.:10
Specifikationer
Om denna produkt
Non corrosive trapezoid blade for use with safety knives.
- Deep-edged, 2-sided ground-edge
- Double bevel edge-ground
- 2-times usable blade
Mountable in:
SECUPRO MAXISAFE
SECUPRO MEGASAFE
SECUPRO 625
SECUNORM MULTISET
SECUNORM PROFI25 MDP
SECUNORM PROFI25