Blades for safety knives, No. 60099
Katalog # 223-0112
Leverantör: MARTOR ARGENTAX
Specifikationer
- Beskrivning:Spare trapezoid blade (no. 60099), two-sided deep edge, rounded tips
- L×B:55,5×19 mm
- Tjocklek:0,63 mm
- Förp.:1 SET
Specifikationer
Om denna produkt
Trapezoid blade with rounded tips for use with safety knives.
- Deep-edged, 2-sided ground-edge
- Double bevel edge-ground
- 2-times usable blade
Mountable in:
SECUPRO MAXISAFE
SECUPRO MEGASAFE
SECUPRO 625
SECUNORM MULTISET
SECUNORM PROFI25