Gateway, Profibus
Gateway 3E (external, for Pilot ONE), 90 – 260V, Profibus
Katalog # HUBE10503
Leverantör: HUBER
Gå till:
Product Details & Documents
This gateway enables the connection of temperature control units to Profibus systems.
Offers a range of possibilities for data communication within SPS and process control systems.
Beställningsinformation: Includes a mounting bracket and connection cable.
Specifikationer
- Modell:Profibus
- Beskrivning:Gateway 3E (external, for Pilot ONE), 90 – 260V