
(-)-Brucine for resolution of racemates, för syntes, Sigma-Aldrich®
Catalog # 8.18308.0050
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:175 - 178 °C
- Molar Mass:394.47 g/mol
- Bulk Density:230 kg/m3
- Storage temperature:+15...+25 °C
- Specific rotation:-86 to -80 °
- Identity (IR):Passes test
Specification Test Results
About this item
:
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:394.47 g/mol
EINECS:206-614-7
UN:1570
:GHS06
Storage Temperature:Omgivande
CAS Number:357-57-3
Formula:C₂₃H₂₆N₂O₄