Order Entry
Sweden
ContactUsLinkComponent
Anti-ATP5B Rabbit Polyclonal Antibody
Anti-ATP5B Rabbit Polyclonal Antibody
Catalog # BOSSBS-8600R
Supplier:  Bioss
Anti-ATP5B Rabbit Polyclonal Antibody
Catalog # BOSSBS-8600R
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
  • Antigensymbol:
    ATP5B
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonymer:
    ATPSB|ATPMB|HEL-S-271
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Rening:
    Purified by Protein A.
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifications

About this item

Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. F1 consists of five distinct subunits, designated ATP5A, ATP5B, ATP5C1, ATP5D and ATP5E, while F0 consists of ten subunits, designated ATP5H, ATP5G1, ATP5I, ATP5G2, ATP5J2, ATP5J, ATP5G3, ATP5S, ATP5F1 and ATP5L. ATP5B, also designated ATPMB, ATPSB or mitochondrial ATP synthetase, beta subunit, is a 529 amino acid protein that localizes to the mitochondrial membrane and exists as a subunit of the F0 complex. ATP5B is encoded by a nuclear gene and assembled with the other subunits encoded by both mitochondrial and nuclear genes. The ATP5B gene is activated by members of the Ets family of transcription factors, suggesting that Ets transcription factors are involved in the enhanced expression of the ATP5B gene in highly proliferating cells and in the coordinate transcription of nuclear genes for mitochondrial proteins. ATP5B mRNA levels vary among species through transcriptional control with high expression levels in heart, lower levels in skeletal muscle and the lowest levels in liver and kidney.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ATP5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat