Order Entry
Sweden
ContactUsLinkComponent
Anti-KLHL22 Rabbit Polyclonal Antibody
Anti-KLHL22 Rabbit Polyclonal Antibody
Katalog # BOSSBS-7807R
Leverantör:  Bioss
Anti-KLHL22 Rabbit Polyclonal Antibody
Katalog # BOSSBS-7807R
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Kelch-like 22
  • Antigensymbol:
    KLHL22
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    84861
  • Antigen synonymer:
    2610318I18Rik.|Klhl22|KLH22_HUMAN|RGD1306288|KELCHL|Kelch-like protein 22
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Rening:
    Purified by Protein A.
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin ligase complex required for cell division. BCR E3 ubiquitin ligase complexes mediate the ubiquitination of target proteins.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: KLHL22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat