Order Entry
Sweden
ContactUsLinkComponent
Anti-DOCK1 Rabbit Polyclonal Antibody
Anti-DOCK1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7110R
Supplier:  Bioss
Anti-DOCK1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7110R
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Dedicator Of Cytokinesis 1
  • Antigensymbol:
    DOCK1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Gen-ID:
    1793
  • Antigen synonymer:
    DOCK180|ced5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Rening:
    Purified by Protein A.
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifications

About this item

Involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. Functions as a guanine nucleotide exchange factor (GEF), which activates Rac Rho small GTPases by exchanging bound GDP for free GTP. Its GEF activity may be enhanced by ELMO1.Tissue specificity: Highly expressed in placenta, lung, kidney, pancreas and ovary. Expressed at intermediate level in thymus, testes and colon.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat