Order Entry
Sweden
ContactUsLinkComponent
Anti-DEPDC1 Rabbit Polyclonal Antibody
Anti-DEPDC1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6525R
Supplier:  Bioss
Anti-DEPDC1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6525R
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    DEP Domain Containing 1
  • Antigensymbol:
    DEPDC1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    55635
  • Antigen synonymer:
    DEPDC1A|cell cycle control protein SDP35|DEP.8|DEPDC1-V2|DEP domain-containing protein 1A|SDP35|5830484J08Rik
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Rening:
    Purified by Protein A.
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifications

About this item

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DEPDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat