

O-Benzylhydroxylamine för syntes, Sigma-Aldrich®
Catalog # 8.20617.0010
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:10 ml
- Seq:2275
Specifications
Specification Test Results
- Solubility:47 g/l (15 °C)
- Molar Mass:123.16 g/mol
- Boiling Point:49 °C (1 hPa)
- Vapor Pressure:2.09 hPa (58 °C)
- Flash Point:91.5 °C
- Density:1.064 g/cm3 (23 °C)
- pH:8 (47 g/l, H2O)
- Ignition Point:260 °C DIN 51794
- Viscosity Kinematic:3.34 mm2/s (23 °C)
- Kinematic viscosity:3,34 mm2/s (23 °C)
- Assay (GC, area%):≥99% (a/a)
- Identity (IR):Passes test
- Storage temperature:+15...+25 °C
Specification Test Results
About this item
:
:123.155 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:2735
:GHS05
:GHS07
Storage Temperature:Omgivande
CAS Number:622-33-3
Formula:C₇H₉NO
MDL Number:MFCD00221709