Data Logger, Einstein LabMate
Om denna produkt
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
- Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
- Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
- Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
: Pressure kit must be ordered separately.
2 Alternativ tillgängliga nedan
Product Details & Documents
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
- Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
- Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
- Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
: Pressure kit must be ordered separately.
Specifications for Product Family
Specifikationer
- Catalog No:
- FOURENLMSEN
- FOURENLM
- Gränssnitt:
- USB
- USB
Specifikationer
Product Family Options
Produktinformation
- TillgänglighetPrice
- FOURENLMSENData logger for science and education, einstein™ LabMate+Data logger for science and education, einstein™ LabMate+Log in för att kolla tillgänglighet och ert pris1 * 1 ItemsSpecifikationer:
More
Specifications
Cat. No.FOURENLMSENGränssnittUSB - FOURENLMData logger for science and education, einstein™ LabMateData logger for science and education, einstein™ LabMateLog in för att kolla tillgänglighet och ert pris1 * 1 ItemsSpecifikationer:
More
Specifications
Cat. No.FOURENLMGränssnittUSB
Accessories
Produktinformation
- Katalog #TillgänglighetPrice
- Sensor adapter
Produktinformation
- Katalog #TillgänglighetPrice
- Blood pressure sensor for einstein™ LabMateBlood pressure sensor for einstein™ LabMateFOURENBLD098Log in för att kolla tillgänglighet och ert pris1 * 1 Items
Produktinformation
- Katalog #TillgänglighetPrice
- Sensor cable