Orders LinkContactUsLinkComponent
Tablet, einstein tablet
Tablet, einstein tablet
Leverantör:   FOURIER SYSTEMS
Tablet, einstein tablet
Leverantör:  FOURIER SYSTEMS
Certificates
Tablet, einstein tablet
2 Alternativ tillgängliga nedan
Product Details & Documents
The einstein ™ Tablet + is a full Android tablet with a built-in science lab designed for primary and secondary science, technology engineering and mathematics education (K-12 STEM).
  • Contains 8 built-in sensors, commonly used in most science curricula, and connects to up to 8 external sensors
  • Designed to be used both in the science lab and also in a broader educational environment
  • Free apps and software included
Built-in sensors include:
Temperature
Humidity
Microphone
Light
UV
Heart rate - jack for external sensor
GPS
Accelerometer
Product Family Options
Produktinformation
  • Tillgänglighet
    Price
  • FOURENTAB
    einstein™ Tablet
    einstein™ Tablet
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items
    1 * 1 Items
  • FOURENTABSEN
    einstein™ Tablet +
    einstein™ Tablet +