Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lock
Lock, flaskor
Pouring rings for square laboratory bottles, DURAN®
Pouring rings for square laboratory bottles, DURAN®
Pouring rings for square laboratory bottles, DURAN®
Leverantör:
DWK Life Sciences
Certifikat
Expand image
Pouring rings for square laboratory bottles, DURAN®
Leverantör:
DWK Life Sciences
Om denna produkt
PP, blue.
For DIN GL threads
Heat resistant up to +140 °C
Autoclavable