Halogen Lamp
Halogen lamp, with handle, 500 W
Katalog # BINHU8476713-230
Marketsource
Leverantör: 3B Scientific
Gå till:
Product Details & Documents
Strong light source for indoor experiments with solar cells, as alternative to strong sunlight.
Specifikationer
- Beskrivning:Halogen lamp, with handle
- Effekt (W):500 W