Round process tank
Round tank without drain, in PFA, 4 l
Katalog # ENTEE14-35-01-0215
Leverantör: ENTEGRIS
Gå till:
Produktdetaljer och dokument
Injection molded round tank, in PFA material.
- Tanks should be properly supported in heated applications
- Maximum recommended temperature limit for PFA tanks is 180 °C
Specifikationer
- Kapacitet:4 l
- Beskrivning:Round tank without drain, in PFA
Accessories
Produktinformation
- Katalog #TillgänglighetPrice
- Cover for round tank, in PFACover for round tank, in PFAENTEE14-35-02-0215Log in för att kolla tillgänglighet och ert pris1 * 1 Items