Natural magnetic ore
Catalog # BINHU19557
Supplier: 3B Scientific
Specifications
- Beskrivning:Natural magnet
- Förp.:1
Specifications
About this item
Unfinished, walnut sized stone made of magnetic iron ore (magnetite).
Unfinished, walnut sized stone made of magnetic iron ore (magnetite).