Diffraction gratings
Marketsource
Leverantör: 3B Scientific
Gå till:
Om denna produkt
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
3 Alternativ tillgängliga nedan
Product Details & Documents
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
Product Family Options
Produktinformation
- TillgänglighetPrice
- BINHU14100Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mmDiaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mmLog in för att kolla tillgänglighet och ert pris1 * 1 ItemsSpecifikationer:MarketsourceMarketsource
- BINHU14101Diaphragm with 4 double slits, Slit spacing: 0,15 mm, Slit width: 0,25 mm; 0,50 mm; 0,75 mm; 1,00 mmDiaphragm with 4 double slits, Slit spacing: 0,15 mm, Slit width: 0,25 mm; 0,50 mm; 0,75 mm; 1,00 mmLog in för att kolla tillgänglighet och ert pris1 * 1 ItemsSpecifikationer:MarketsourceMarketsource
- BINHU14102Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mmDiaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mmLog in för att kolla tillgänglighet och ert pris1 * 1 ItemsSpecifikationer:MarketsourceMarketsource