Insect prepared slides
Catalog # LIED4300
Supplier: LIEDER JOHANNES
Specifications
- Beskrivning:Insecta prepared slides, elementary set incl. 25 slides
- Förp.:1 SET
Specifications
About this item
Transverse section (t.s.).
- Musca domestica, housefly, leaking-sucking mouth parts
- Pieris, butterfly, sucking mouth parts
- Carabus, ground beetle, biting mouth parts (carnivore)
- Melolontha, cockchafer, chewing mouth parts (herbivore)
- Pyrrhocoris, bug, piercing sucking mouth parts
- Bombyx mori. silkworm moth, chewing mouth parts
- Apis mellifica, honey bee, leaking sucking mouth parts of worker
- Culex pipiens, mosquito, piercing sucking mouth parts
- Melolontha, cockchafer, antenna with sense organs
- Bombyx mori, silkworm moth, feathered antenna
- Apis mellifica, anterior leg with eye brush
- Apis mellifica, posterior leg with pollen basket
- Musca domestica, house fly, leg with pulvilli
- Apis mellifica, anterior and posterior wings
- Pieris, butterfly, portion of wings with scales
- Trachea from insect
- Spiracle from insect
- Cornea isolated from insect eye
- Apis mellifica, honey bee, sting and poison sac
- Apis mellifica, head with compound eyes and brain (t.s)
- Bombyx mori, silkworm (t.s) showing silk spinning glands
- Drosophila, fruit fly of adult
- Ctenocephalus canis, dog flea of an adult
- Culex pipiens, mosquito, larva
- Chironomus, gnat, larva
Ordering information: Includes 25 microscopic slides with accompanying brochure.