Laboratory washer-Disinfector, PG 8527
Leverantör: MIELE
Certifikat
Om denna produkt
The washer-disinfector PG 8527 offers great added value for central and decentralised processing of large volumes of instruments: greater cleaning capacity, greater process reliability and greater economy.
- New Profitronic controller with PerfectTouch control
- Patented maintenance-free conductivity meter
- Improved hygienic wash chamber with a unique spray arm monitoring system
- Highly accurate detergent dispensing monitor using ultrasound technology
- Improved HEPA-filtered forced air drying system
- Full-view glass door with chamber lighting
Construction: Freestanding unit with single lift-up door, the frame is in AISI 304 stainless steel, external casing in AISI 304 stainless steel, grain brushed to 220 grade.
Comprehensive safety features in accordance with EN ISO 15883
Certifications: Comprehensive safety features in accordance with EN ISO 15883
Ordering information: Baskets and inserts must be ordered separately.