Avantor® ACE® CN, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
These Avantor® ACE® CN columns provide great reproducibility and column lifetime.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Can be used under both reversed phase and normal phase conditions
- Capillary to preparative dimensions
- Wide range of particle sizes (2 µm to 10 µm) and complementary phases
- Useful for the separation of polar analytes and compounds with double and triple bonds
- Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Specifications
- Catalog No:
- ACE-124-2046
- ACE-124-3004
- ACE-124-3046
- ACE-124-1546NP
- ACE-124-0503
- ACE-124-1220
- ACE-124-1246
- ACE-124-0502
- ACE-124-0303
- ACE-124-0103GD
- ACE-124-2530
- ACE-124-1204
- ACE-124-2546
- ACE-124-2510
- ACE-124-2520
- ACE-124-0520
- ACE-124-0530
- ACE-124-3039
- ACE-124-1502
- ACE-124-7530
- ACE-124-3030
- ACE-124-2504
- ACE-124-3020
- ACE-124-1530
- ACE-124-1546
- ACE-124-1510
- ACE-124-1520
- ACE-124-1002
- ACE-124-1504
- ACE-124-0546
- ACE-124-7520
- ACETACE-124-1539
- ACE-124-1046
- ACE-124-7503
- ACE-124-2503
- ACE-124-1020
- ACE-124-1030
- Kolbelastning:
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- Endcapped:
- Ja
- Ja
- Ja
- Ja
- Ja
- Yes
- Ja
- Yes
- Ja
- Yes
- Ja
- Ja
- Ja
- Ja
- Yes
- Yes
- Ja
- Ja
- Yes
- Yes
- Ja
- Yes
- Yes
- Ja
- Yes
- Ja
- Yes
- Ja
- Ja
- Yes
- Ja
- Ja
- Ja
- Ja
- Yes
- Yes
- Max. tryck:
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- 276 bar
- pH område:
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10