UHPLC columns, Ultra C18 (USP L1)
Leverantör: Restek
Certifikat
Om denna produkt
Ultra C18 is a general purpose reversed phase column with retentive and high-purity packing that exhibits excellent peak shape for a wide range of compounds.
Specifikationer
- Catalog No:
- REST9174315
- Kolbelastning:
- 20%
- Endcapped:
- Ja, Ja
- pH område:
- 2,5 - 8
- Temperature limit:
- 80 °C
- USP code:
- L1, L1