Överföringsvagn
Catalog # 141-0572
Supplier: MIELE
Specifications
- Beskrivning:MF 27/28-1 Transfer cart for glassware washing machine, Miele G 8527/28
- B×D×H:740×930×1050 mm
- Förp.:1
- Antal hyllor:1
Specifications
About this item
For loading and unloading washers with backets and inserts.
- With removable drip tray (W×D×H: 603×866×70 mm)
- Both ends can dock onto machine, tables, sluices or conveyors
- Docking height 850, –100, + 150 mm
- 4 lockable wheels, Ø 100 mm