Wet film thickness gauges
Supplier: Industrial Physics
Certificates
About this item
Hexagonal/octagonal precision measuring comb made of heavy stainless steel - The high-grade stainless steel will not be affected by acid or base elements.
- Available in microns and mils scale
- For decorative paints and primers (20 to 370 µm)
- For protective coatings and coating with high solids content (25 to 2000 µm)
- For extremely thick films such as floor coating, fillers, fire proofing, plasters and adhesives (50 to 10000 µm)
- For protective coatings and coating with high solids content (1 to 80 mils)
Specifications
- Catalog No:
- SIMESP4020
- SIMESP4000
- SIMESP4060
- SIMESP4010
- Modell:
- WG-3
- WG-1
- WG-2
- Noggrannhet:
- 60 μm
- 10 μm
- 30 μm
- Lagrade resultat:
- 71
- 24
- 36