Avantor® ACE® C18-HL, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® C18-HL columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3 to 15 µm) and complementary phases
- Especially suited for preparative work due to higher carbon load
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Delivery: Method validation kits contain three different batches of the same phase.
Specifications
- Catalog No:
- ACE-321-1046
- ACE-321-1020
- ACE-321-1030
- ACE-321-1010
- ACE-321-1003
- ACE-321-1546
- ACE-321-1520
- ACE-321-7520
- ACE-321-7530
- ACE-321-1530
- ACE-321-1510
- ACE-321-1503
- ACE-321-0530
- ACE-321-0546
- ACE-321-0120GD
- ACE-321-2503
- ACE-321-2504
- ACE-321-0510
- ACE-321-1220
- ACE-321-1203
- ACE-321-0346
- ACE-321-3020
- ACE-321-2546
- ACE-321-3046
- ACE-321-0110GD
- ACE-321-2520
- ACE-321-3004
- ACE-321-0103GD
- ACE-321-2530
- ACE-321-3030
- ACE-321-0520
- ACE-321-2510
- Kolbelastning:
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- 20,0%
- Endcapped:
- Ja
- Yes
- Yes
- Yes
- Ja
- Ja
- Yes
- Ja
- Yes
- Yes
- Ja
- Ja
- Yes
- Ja
- Ja
- Ja
- Yes
- Yes
- Ja
- Ja
- Yes
- Ja
- Ja
- Ja
- Ja
- Ja
- Yes
- Yes
- Ja
- Max. tryck:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH område:
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1