Peqlab peqGOLD, Agarose, Universal
Catalog # 732-2789
Supplier: VWR Chemicals
Specifications
- Beskrivning:Universal agarose
- Förp.:500 g
Specifications
About this item
High quality standard agarose for routine analysis with broad separation range (0,05 to 50 kbp). Ideal for blotting applications due to high gel strength. Free from RNases, DNases and enzyme inhibitors.