Gas detection tubes for chlorine
Leverantör: Dräger
Certifikat
Om denna produkt
To measure the 'spot concentrations' of chlorine for use with an Accuro® pump.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
Cross sensitivity: Bromine is indicated with the same sensitivity. Chlorine dioxide and nitrogen dioxide are indicated as well, but with different sensitivities.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Specifikationer
- Catalog No:
- 151-0191
- 151-0195
- Modell:
- Chlorine 50/a
- Chlorine 0.2/a
- Färgförändring:
- Green grey to orange brown
- White to yellow orange
- Fuktighet:
- ≤ 40 mg/l H₂O
- ≤ 15 mg/l H₂O
- Mättid:
- 20 seconds
- 3 minutes/20 seconds
- No. of strokes:
- 1
- 10/1
- Standard deviation:
- ±10 - 15 %
- ±10 - 15 %
- Standard range:
- 50 - 500 ppm
- 0,2 - 3/3 - 30 ppm
- Temperaturområde:
- 10…40 °C
- 10…40 °C