Guaiacol detection kit
Katalog # 710-0751
Leverantör: DOEHLER GROUP
CAS-nummer:
Specifikationer
- Förpackningstyp:Bottle
- Förp.:40 Tests
- Standard:IFU Method No. 12
- Beskrivning:Guaiacol-detektionskit
- Fyllning:-
Specifikationer
Om denna produkt
Enzymatic test kit for the detection of off-flavour forming Alicyclobacillus. Guaiacol Detection Kit is a reliable and sensitive enzymatic detection of Guaiacol-forming Alicyclobacilli. When Guaiacol-forming bacteria are present, the transparent to slightly yellow Guaiacol is transformed into the clearly-visible, brownish complex Tetraguaiacol with the enzyme peroxidase from the Detection Kit.
- Comprehensive Detection Kit including BAT Broth in tubes and Guaiacol Standard Solution
- Simple visual analysis without additional technical equipment