Suture practice leg
Produktdetaljer och dokument
Made with a soft vinyl skin over a core of stitchable foam to provide a lifelike suturing experience for students or a realistic suturing demonstration by an instructor. Realistic skin texture with wrinkles and pores. Soft and pliable for easy sewing, the skin is tough enough that sutures will not pull out when tightened. Each unit provides three 'wounds'. These 'wounds' can be sutured repeatedly until the skin around them is finally worn out; then, new 'wounds' can be cut. Although disposable, the trainers provides hundreds of suturing experiences before wearing out. Conservatively, over one hundred cuts can be made on the trainer and each of these cuts can be sutured several times.
- Complete with a starter suturing kit and instruction booklet
- Dimensions: 62×18×13 cm
Specifikationer
- Beskrivning:Suture Practice Leg