Synapse
Synapse
Katalog # BINH1000237
Marketsource
Leverantör: 3B Scientific
Gå till:
Produktdetaljer och dokument
This synapse model is a great addition to any anatomical lesson of the human nervous system. The synapse model features the endoplasmic reticulum, mitochondria and the membranes of the synaptic gap. Also depicted by the synapses model are 5 smaller relief models of the main synapse variations.
- A great addition to any anatomical lesson of the human nervous system
Specifikationer
- Beskrivning:Synapse