Peqlab peqGOLD, Protein Marker II (Unstained)
Katalog # 27-2010
Leverantör: VWR Chemicals
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:Protein Marker II (unstained, ready to use), 2×250 µl, sufficient for 50 lanes
Specifikationer
Om denna produkt
Protein Marker II for accurate molecular weight determination of proteins by SDS-PAGE.
- Mix of 14 recombinant proteins
- Range: 10 to 200 kDa
- Each protein with Strep-Tag® II sequence (WSHPQFEK)
- Highlighted 50 kDa band for easy orientation
- Ready to use for a fast, convenient workflow