Peqlab peqGOLD, DNA Ladder, 1 kb
Catalog # 25-2030
Supplier: VWR Chemicals
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:DNA ladder, 1 kb (concentrated), 500 ng/µl, 5×100 µl, sufficient for 250 lanes
Specifications
About this item
DNA ladder for quantitative and qualitative analysis of dsDNA.
- Range: 14 fragments, 250 to 10000 bp in size
- Three highlighted bands for easy orientation
- Also available as ready to use mix containing Orange G dye instead of bromophenol blue