Qualitative filter papers, technical, smooth, grade 6
Supplier: Sartorius
Certificates
About this item
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Fast rate of filtration, high wet strength. All-purpose filter papers; also recommended for malt and beer analysis or degassing; also suitable for coarser or gelatinous precipitates; for clarification of dried sugar beet shreds in sugar factories; essences; spirits.
Specifications
- Catalog No:
- SARTFT-3-312-385
- SARTFT-4-312-210
- SARTFT-3-312-320
- SARTFT-3-312-055
- SARTFT-3-312-270
- SARTFT-3-312-240
- SARTFT-4-312-385
- SARTFT-4-312-270
- SARTFT-3-312-125
- SARTFT-3-312-210
- 515-1578
- SARTFT-4-312-125
- SARTFT-4-312-110
- SARTFT-4-312-090
- SARTFT-4-312-150
- SARTFT-3-312-185
- SARTFT-3-312-070
- 512-0534
- SARTFT-3-312-150
- SARTFT-3-312-110
- SARTFT-4-312-185
- SARTFT-3-312-090
- SARTFT-4-312-240
- Klass:
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- Vikt:
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- Tjocklek:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- Partikelretention:
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- 10 - 13 µm
- Filtreringshastighet:
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- 15 s
- Sprängtryck i vått tillstånd:
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa
- 30 kPa