Glass microfiber filters with organic binder, grade MN 85/90
Supplier: MACHEREY-NAGEL
Certificates
About this item
The MN 85/90 grade is the most widely used, general purpose glass fiber filter with very good mechanical stability. The filter contains an organic binder.
- High mechanical stability
- General purpose filter
- Stable up to 200 °C
Specifications
- Catalog No:
- 515-9798
- 515-9797
- MANA405015
- MANA405027
- MANA405005
- MANA4050025
- 515-9795
- MANA4050037
- MANA4050045
- MANA405024
- MANA405007
- 515-9796
- Vikt:
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- Tjocklek:
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- 0,40 mm
- Partikelretention:
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- 0,5 μm
- Filtreringshastighet:
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- 15 s (air)
- Max. temperature:
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C