Peqlab peqGOLD, DNA Ladders, 50 bp
Katalog # 25-2000
Leverantör: VWR Chemicals
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:DNA ladder, 50 bp (concentrated), 500 ng/µl, 1×100 µl, sufficient for 50 lanes
Specifikationer
Om denna produkt
DNA ladder for quantitative and qualitative analysis of dsDNA.
- Range: 13 distinct fragments 50 to 1000 bp in size
- Two highlighted bands for easy orientation
- Also available as ready to use mix containing Orange G dye instead of bromophenol blue