Jig trays
Catalog # ENTEP44-JIG-101-4
Supplier: ENTEGRIS
Specifications
- Beskrivning:Standard, carbon fiber-filled PES
- Förp.:5
Specifications
About this item
Patented JEDEC thick profile jig tray.
- Holds three H44 or P44 trays
- Maximum temperature: 180 °C
- Order in multiples of five trays