Plastic cuvette for LICO
Katalog # 705-1169
Leverantör: Hach
Specifikationer
- Skiktdjup:50 mm
- Material:Plast
- Beskrivning:Spectrophotometer cuvettes
- Höjd:10 mm
- Förp.:50
Specifikationer
Om denna produkt
Disposable rectangular cuvette for spectral measurements of liquids.