VWR® Gold-N-Gel™, RNA dye
Catalog # N734-5ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Förp.:5 mL
- Beskrivning:RNA dye (200X)
- Seq:01
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
About this item
Gold-N-Gel RNA dye is compatible with standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illumination with UV light. A standard ethidium bromide filter can be used for gel documentation.
- Non mutagenic, fluorescent in-gel stain for immediate visualisation of RNA bands on denaturing agarose gels
- Immediate band visualisation with UV illumination - No post-run staining or destaining
- Easy-to-use: Simply add to melted agarose prior to casting
Delivery: Supplied as a 200X concentrate.