VWR® EZ-Vision®, DNA Ladder, 1 kb
Catalog # N854-1.2ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Förp.:1,2 mL
- Beskrivning:EZ-Vision® 1 kb DNA ladder
- Seq:02
Specifications
About this item
EZ-Vision® 1 kb DNA ladder is a pre-mixture of DNA marker, loading buffer and a proprietary fluorescent DNA visualisation dye that can be applied directly to the gel. This non toxic, non mutagenic dye eliminates the need for ethidium bromide staining. The 1 kb ladder consists of 10 double-stranded bands with molecular weights between 200 and 10 000 bp. The intensity of individual bands varies to facilitate easy identification.
- Instant visualisation
- 'Load-and-go' formula makes it easy and convenient
- Eliminates the use of ethidium bromide