VWR® EZ-Vision®, DNA Ladder, 100 bp
Katalog # N856-300UL
Leverantör: VWR Chemicals
CAS-nummer:
Specifikationer
- Förp.:300 µl
- Beskrivning:EZ-Vision® 100 bb DNA ladder, sufficient for 50 assays
- Environmentally Preferable:Safer Human and Environmental Health
Specifikationer
Om denna produkt
EZ-Vision® 100 bp DNA ladder is a pre-stained molecular weight ladder in loading buffer that allows immediate visualisation of DNA ladder post-electrophoresis.
- 13 fragments ranging from 100 to 3000 bp with reference bands for easy identification
- 'Load-and-go' formula with instant visualisation
- Eliminates the use of ethidium bromide