Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CACNA1G Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S178A-9]
Catalog # ABNOMAB11563
Supplier:  Abnova
Anti-CACNA1G Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S178A-9]
Catalog # ABNOMAB11563
Supplier:  Abnova

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Calcium Channel, Voltage-dependent, T type, alpha 1G subunit
  • Antigensymbol:
    CACNA1G
  • Klonalitet:
    Monoclonal
  • Klon:
    S178A-9
  • Konjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG1
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Korsad adsorption:
    No
  • Mått:
    Liquid
  • Gen-ID:
    12291
  • Antigen synonymer:
    Cav3.1d|[a]1G|mKIAA1123|a1G|alpha-1G
  • Aminosyretal:
    2052 to 2172
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol)
  • Förvaringstemperatur:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g.
  • Rening:
    Protein G Purified
  • Storlek:
    100 μg
  • Förp.:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant of Cacna1g.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: