Glass Beads, SiLibeads®
LINR7503-20KG
- Ø:0,75 - 1,00 mm
- Beskrivning:Glass beads
- Förp.:20 kg
Aluminium-borosilicate glass.
SiLibeads® Type SL glass beads used as grinding media for horizontal and vertical bead mills.
Aluminium-borosilicate glass.
SiLibeads® Type SL glass beads used as grinding media for horizontal and vertical bead mills.