Order Entry
Sweden
ContactUsLinkComponent
Anti-FFAR4 Rabbit Polyclonal Antibody
Anti-FFAR4 Rabbit Polyclonal Antibody
Katalog # CAYM14265-1
Leverantör:  Cayman Chemical
Anti-FFAR4 Rabbit Polyclonal Antibody
Katalog # CAYM14265-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    FFAR4
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Korsad adsorption:
    No
  • Aminosyretal:
    364 to 375
  • Storage buffer:
    TBS, pH 7,4
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human GPR120 amino acids 364-375
  • Rening:
    purified by affinity chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: FFAR4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human