Order Entry
Sweden
ContactUsLinkComponent
Anti-MeCP2 Rabbit Polyclonal Antibody
Anti-MeCP2 Rabbit Polyclonal Antibody
Catalog # CAYM13775-1
Leverantör:  Cayman Chemical
Anti-MeCP2 Rabbit Polyclonal Antibody
Catalog # CAYM13775-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    MECP2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Aminosyretal:
    11 to 25, 181 to 195
  • Storage buffer:
    PBS containing 0,2% gelatin and 0,05% sodium azide
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    a mixture of synthetic peptides corresponding two amino acids 268-282 and 337-352 of human MBD4
  • Rening:
    purified by Protein G chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: MeCP2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human