ContactUsLinkComponent
Anti-TLR4 Mouse Monoclonal Antibody [clone: HTA125]
Anti-TLR4 Mouse Monoclonal Antibody [clone: HTA125]
Catalog # CAYM13589-1
Supplier:  Cayman Chemical
Anti-TLR4 Mouse Monoclonal Antibody [clone: HTA125]
Catalog # CAYM13589-1
Supplier:  Cayman Chemical
Specifications
  • Antikroppstyp:
    Primär
  • Antigensymbol:
    TLR4
  • Klonalitet:
    Monoclonal
  • Klon:
    HTA125
  • Konjugation:
    Unconjugated
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Immunutfällning:
    Yes
  • Isotyp:
    IgG2a kappa
  • Korsad adsorption:
    No
  • Neutralisation:
    Yes
  • Storage buffer:
    PBS containing 0,05% BSA and 0,05% sodium azide
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Ba/F3 cell line expressing human TLR4 cell surface antigen
  • Rening:
    purified by Protein G chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1
Specifications
About this item
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: HTA125
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: