Order Entry
Sweden
ContactUsLinkComponent
Anti-TLR12 Rabbit Polyclonal Antibody
Anti-TLR12 Rabbit Polyclonal Antibody
Katalog # CAYM13586-1
Leverantör:  Cayman Chemical
Anti-TLR12 Rabbit Polyclonal Antibody
Katalog # CAYM13586-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    TLR12
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Flödescytometri:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Aminosyretal:
    911 to 926
  • Storage buffer:
    PBS containing 0,2% gelatin adn 0,05% sodium azide
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    mouse TLR12 amino acids 911-926
  • Rening:
    purified by Protein G chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: TLR12
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat