Anti-Smt3 Mouse Monoclonal Antibody [clone: N/A]
Catalog # CAYM12021-500
Supplier: Cayman Chemical
Specifications
- Antikroppstyp:Primär
- Antigensymbol:Smt3
- Klonalitet:Monoclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- Isotyp:IgG1
- Western blot:Yes
- Korsad adsorption:No
- Aminosyretal:1 to 97
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant yeast Smt3 amino acids 1-97
- Rening:purified by Protein G chromatography
- Storlek:500 µg
- Förp.:500 µG
Specifications
About this item
Type: Primary
Antigen: Smt3
Clonality: Monoclonal
Clone: N/A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: