Anti-BLT1 Rabbit Polyclonal Antibody
Catalog # CAYM120114-500
Leverantör: Cayman Chemical
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:BLT1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Bovine,Mouse
- Western blot:Yes
- Korsad adsorption:No
- Aminosyretal:331 to 352
- Storage buffer:TBS, pH 7,4, containing 50% glycerol, 0,5% BSA, and 0,02% sodium azide
- Sequence:ALEPGPSESLTASSPLKLNELN
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human BLT1 receptor amino acids 331-352
- Storlek:500 µl
- Förp.:500 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: BLT1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse