Anti-ENHO Rabbit Polyclonal Antibody
CAYM10381-1
- Antikroppstyp:Primär
- Antigensymbol:ENHO
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Aminosyretal:34 to 76
- Storage buffer:TBS, pH 7,4
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human adropin amino acids 34-76
- Rening:purified by affinity chromatography
- Storlek:1 Ea
- Förp.:1
Type: Primary
Antigen: ENHO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human