Order Entry
Sweden
ContactUsLinkComponent
Anti-IP6K2 Mouse Monoclonal Antibody [clone: 4F10]
Anti-IP6K2 Mouse Monoclonal Antibody [clone: 4F10]
Katalog # CAYM10239-1
Leverantör:  Cayman Chemical
Anti-IP6K2 Mouse Monoclonal Antibody [clone: 4F10]
Katalog # CAYM10239-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    IP6K2
  • Klonalitet:
    Monoclonal
  • Klon:
    4F10
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG2b kappa
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Storage buffer:
    TBS, pH 7,4
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human IP6K2
  • Rening:
    purified by Protein A chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: IP6K2
Clonality: Monoclonal
Clone: 4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse