Order Entry
Sweden
ContactUsLinkComponent
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Catalog # CAYM10009763-1
Supplier:  Cayman Chemical
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Catalog # CAYM10009763-1
Supplier:  Cayman Chemical

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    PGR
  • Klonalitet:
    Monoclonal
  • Klon:
    608
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Modifiering:
    phospho-Ser294
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser294 of human progesterone receptor
  • Rening:
    purified by affinity chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifications

About this item

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 608
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human